Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Safety wash bottles, narrow neck, printed, Nalgene®

Supplier: Thermo Scientific
Nalgene®
LDPE, translucent except sodium hypochlorite (white LDPE with light protection) and acetone (PPCO).

  • Good chemical resistance, heat resistant up to +80 °C (for short periods up to +90 °C)
  • Non metallic vent valve keeps contents in the bottle, no involuntary dripping
  • Coloured contents label with the chemical's name, ICS formula, CE index number and CAS number
  • GHS compliant symbols

With PP spray caps except sodium hypochlorite (HDPE) and coloured contents label. With hazard symbols and descriptions of hazards as required by the Hazardous Substance Ordinance. To allow easy hazard and chemical identification, as well as to prevent cross-contamination, each bottle has a colour bar that corresponds to its cap colour.

Ordering information: Assortment includes bottles (500 ml) for all chemicals listed except sodium hypochlorite.



 

ORDER

Expand All / Collapse All
Imprint Capacity Colour code Pk VWR Catalog Number Quantity
Acetone 500 ml Red 6 215-0159
Ethanol 500 ml White 6 215-0172
Isopropanol 500 ml Yellow 6 215-0174
Methanol 500 ml Green 6 215-0173
Sodium hypochlorite 500 ml White 6 215-0177
Distilled water 500 ml Natural 6 215-0176
Assortment 500 ml 5 215-0124
Ethanol 1000 ml White 4 215-0178
Isopropanol 1000 ml Yellow 4 215-0183
Methanol 1000 ml Green 4 215-0179
Distilled water 1000 ml Natural 4 215-0184
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping