Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Waste bins

Supplier: MSH
PP

These containers with two side handles and one handle on the lid are suitable for the collection and storage of sharp pointed or cutting objects, and infectious medical waste.

  • Perforation resistant
  • Hot melt adhesive joint forms an impenetrable seal
  • Base moulded to the shape of the lid for extra stability and easy stacking

UN approved and with ADR approval, conforms to the norm NF X 30 - 505



 

ORDER

Expand All / Collapse All
Description Colour Capacity W×D×H Pk VWR Catalog Number Quantity
Admissible weight 15 kg - 30 l 400×300×350 mm 1 119-0403
Admissible weight 25 kg - 50 l 400×300×500 mm 1 119-0408
Admissible weight 25 kg - 60 l 400×305×575 mm 1 129-1800
Admissible weight 25 kg Blue 50 l 400×300×500 mm 1 HUARCON-50R/BLUE
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty