Hippuric acid
Supplier: Molekula
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Benzoylaminoacetic acid
, N-Benzoylglycine
, Benzoyl amidoacetic acid
, Bz-Gly-OH
, Benzoyl glycocoll
, 2-Benzamidoacetic acid
MOLE24771195-1KGEA
0
GBP
MOLE24771195-1KG
Hippuric acid
Hippuric acid
Formula:
C₆H₅C(O)NHCH₂COOH MW: 179.18 g/mol Boiling Pt: 399 °C (1013 hPa) Melting Pt: 271 °C Density: 1,308 g/cm³ (20 °C) |
MDL Number:
MFCD00002692 CAS Number: 495-69-2 EINECS: 207-806-3 Merck Index: 13,04735 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...