Anti-ARMCX1 Rabbit Polyclonal Antibody
ANTIA101214-100
New Product
- Antibody type:Primary
- Antigen name:AD032
- Antigen symbol:ARMCX1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# Q9P291
- Antigen synonyms:ARMCX 1|Armadillo repeat containing X linked 1|ARMX1_HUMAN|ALEX1|ARM protein lost in epithelial cancers on chromosome X 1|Armadillo repeat-containing X-linked protein 1|Armadillo repeat containing X linked protein 1|ARMCX1|Arm protein lost in epithelial cancers X chromosome 1
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:49 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human ARMX1 (amino acids 1-50)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Pack type:Plastic vial
- Pk:100 µG
Rabbit polyclonal antibody to ARMX1 for WB and ELISA with samples derived from human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:20000
Type: Primary
Antigen: ARMCX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human