Anti-AMACR Rabbit Recombinant Antibody [clone: AMACR/3931R]
ANTIA251798-100
New Product
- Antibody type:Primary
- Antigen name:2 arylpropionyl CoA epimerase
- Antigen symbol:AMACR
- Clonality:Recombinant
- Clone:AMACR/3931R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# Q9UHK6
- Antigen synonyms:Alpha methylacyl-CoA racemase deficiency, included|2-methylacyl-CoA racemase|Alpha methylacyl CoA racemase|Alpha-methylacyl-CoA racemase|AMACR|Alpha methylacyl Coenzyme A racemase|AMACR_HUMAN|AMACR deficiency, included|2 methylacyl CoA racemase
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:42 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human AMACR protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Recombinant rabbit monoclonal [AMACR/3931R] antibody to AMACR for WB and IHC-P with samples derived from Human.
- Validated applications: WB, IHC-P
- Recommended dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: AMACR
Clonality: Recombinant
Clone: AMACR/3931R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human