Anti-Cyclin D1 Mouse Monoclonal Antibody [clone: CCND1/3548]
ANTIA249860-100
New Product
- Antibody type:Primary
- Antigen name:AI327039
- Antigen symbol:Cyclin D1
- Clonality:Monoclonal
- Clone:CCND1/3548
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P24385
- Antigen synonyms:BCL1|B cell leukemia 1|BCL-1 oncogene|B-cell lymphoma 1 protein|B cell CLL/lymphoma 1|B cell lymphoma 1 protein|BCL1 oncogene|BCL-1|BCL 1
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:36 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 115-270 protein, of human CCND1 protein. The exact sequence is proprietary.
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [CCND1/3548] antibody to Cyclin D1 for ELISA with samples derived from human.
- Validated applications: ELISA
Type: Primary
Antigen: Cyclin D1
Clonality: Monoclonal
Clone: CCND1/3548
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human