United Kingdom
Order Entry
Anti-CD146 Mouse Monoclonal Antibody [clone: MCAM/3179]
  ANTIA249297-100
New Product
undefined
Anti-CD146 Mouse Monoclonal Antibody [clone: MCAM/3179]
  ANTIA249297-100
New Product
 
£355.00
   
  1 * 100 µG

 

  • Antibody type:
    Primary
  • Antigen name:
    A32 antigen
  • Antigen symbol:
    CD146
  • Clonality:
    Monoclonal
  • Clone:
    MCAM/3179
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2c kappa
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P43121
  • Antigen synonyms:
    Melanoma associated antigen A32|Cell surface glycoprotein P1H12|Mcam|Melanoma adhesion molecule|CD146 antigen|Cell surface glycoprotein MUC18|CD 146|Gicerin|CD146
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    130 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant human MCAM protein
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [MCAM/3179] antibody to CD146 for ELISA with samples derived from human.

  • Validated applications: ELISA

Type: Primary
Antigen: CD146
Clonality: Monoclonal
Clone: MCAM/3179
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2c kappa
Reactivity: Human