United Kingdom
Order Entry
Human Integrin alpha V beta 3 ELISA Kit
Human Integrin alpha V beta 3 ELISA Kit
  ANTIA74300-96
New Product
 :  
undefined
Human Integrin alpha V beta 3 ELISA Kit
  ANTIA74300-96
New Product
 :  
 
£425.00
   
  1 * 96 Tests

 

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Host:
  • Primary antibody reactivity:
    Human
  • Target protein:
    Integrin alpha V beta 3
  • Description:
    Human integrin alpha V beta 3 ELISA kit
  • Environmentally Preferable:
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross reactivity:
    Human Integrin alpha V beta 3 ELISA Kit exhibits high specificity and excellent specificity for the detection of human Integrin alpha V beta 3. No significant cross-reactivity or interference between Integrin alpha V beta 3 and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    31,25 - 2000 pg/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    100 μl
  • Sensitivity:
    18,75 pg/ml
  • Regulatory status:
    RUO