Mouse Hepcidin ELISA Kit
ANTIA313455-96
New Product
:
- Pk:96 Tests
- Assay duration:One step
- Assay Type:Sandwich
- Format:Pre-coated
- Host:
- Primary antibody reactivity:Mouse
- Target protein:Hepcidin
- Description:Mouse Hepcidin ELISA kit
- Environmentally Preferable:
- Sample type:Serum, plasma or other biological fluids
- Detection method:Colorimetric
- Time to Results:1 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Detection range:100 - 1600 ng/ml
- Storage temperature:4 °C
- Sample volume:40 μl
- Sensitivity:5,63 ng/ml
- Regulatory status:RUO