Deuterium oxide (99.96% D) for NMR spectroscopy

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Heavy waterWater-D2

87156.0100EA 347 GBP
87156.0100 87156.0025 87156.0011
Deuterium oxide (99.96% D) for NMR spectroscopy
Deuterium oxide
Formula: D₂O
MW: 20 g/mol
Boiling Pt: 101.3 °C (1013 hPa)
Melting Pt: 3.8 °C
Density: 1.1 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00044636
CAS Number: 7789-20-0
EINECS: 232-148-9
Merck Index: 12,02984

Order Now

Specification Test Results

Isotopic enrichment (FT NMR 400 MHz)(D) Min. 99.96 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR