Anti-MED31 Rabbit Polyclonal Antibody
Catalog # PRSI16-264
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Mediator complex subunit 31
- Antigen symbol:MED31
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:51003
- Antigen synonyms:mediator of RNA polymerase II transcription|Soh1|CGI-125|3110004H13Rik|mediator complex subunit SOH1|mediator of RNA polymerase II transcription subunit 31|hSOH1|subunit 31 homolog
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-131 of human MED31 (NP_057144,1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: MED31
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat