Anti-TRIM48 Rabbit Polyclonal Antibody
PRSI56-972
New Product
- Antibody type:Primary
- Antigen name:Tripartite motif-containing protein 48
- Antigen symbol:TRIM48
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Amino acid range = 42-71
- Form:Liquid
- Gene ID:Q8IWZ4
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:24 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This TRIM48 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 42-71 amino acids from the N-terminal region of human TRIM48
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
TRIM48 belongs to the TRIM/RBCC family. It contains one B box-type zinc finger, one B30.2/SPRY domain and one RING-type zinc finger. There are two named isoforms.
- For WB starting dilution is: 1:1000
Type: Primary
Antigen: TRIM48
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human