Anti-FCGR2B Recombinant Antibody [Clone: AT10]
PRSI24-290
New Product
- Antibody type:Primary
- Antigen name:Fc gamma receptor IIb
- Antigen symbol:FCGR2B
- Clonality:Recombinant
- Clone:AT10
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:
- Isotype:IgG1 kappa
- Reactivity:
- Environmentally Preferable:
- Cross adsorption:No
- Form:Liquid
- Gene ID:2213
- Antigen synonyms:FCGR2B|FcgRIIb|Low affinity immunoglobulin gamma Fc region receptor II-b
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Human CD32
- Pack type:Vial
- Pk:0,2 mg
Type: Primary
Antigen: Fc gamma receptor IIb
Clonality: Recombinant
Clone: AT10
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1 kappa
Reactivity: Human