4-Chlorostyrene stabilised, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-Chloro-4-vinylbenzene
Stabilisation:
Stabilised with 4-tert-butylpyrocatechol (TBC)
M-Clarity™ quality level = MQ200
8.02458.0010EA
82
GBP
8.02458.0010
8.02458.9999
4-Chlorostyrene stabilised, Sigma-Aldrich®
4-Chlorostyrene
Formula:
C₈H₇Cl MW: 138.6 g/mol Boiling Pt: 192 °C (1013 hPa) Melting Pt: –15.9 °C Density: 1.155 g/cm³ (20 °C) |
MDL Number:
MFCD00000632 CAS Number: 1073-67-2 EINECS: 214-028-8 |
Specification Test Results
Solubility | (20 °C) insoluble |
Molar Mass | 138.59 g/mol |
Boiling Point | 192 °C |
Flash Point | 68 °C |
Refractive Index | 1.5645 (20 °C, 589 nm) |
Density | 1.09 g/cm3 (20 °C) |
Synonyms | 1-Chloro-4-ethenyl-bezene |
Hill Formula | C8H7Cl |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...