4-Chlorostyrene 99% stabilised
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1-Chloro-4-vinylbenzene
ACRO110090500EA
112
GBP
ACRO110090500
ACRO110090100
4-Chlorostyrene 99% stabilised
4-Chlorostyrene
Formula:
C₈H₇Cl MW: 138.6 g/mol Boiling Pt: 192 °C (1013 hPa) Melting Pt: –16 °C Density: 1.16 g/cm³ Flash Pt: 64 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000632 CAS Number: 1073-67-2 EINECS: 214-028-8 UN: 3082 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5645 to 1.5665 (20°C, 589 nm) |
GC | ≥98.5 % |
Stabilizer | 0.1 % tert.-butylcatechol |
Acidity (CH3COOH) | ≤0.02 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...