2-Chlorostyrene ≥97% stabilised
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1-Chloro-2-vinylbenzene
Stabilisation:
Stabilised with 4-tert-butylpyrocatechol (TBC) 0.10 %
B21551.14EA
109
GBP
B21551.14
2-Chlorostyrene ≥97% stabilised
2-Chlorostyrene
Formula:
C₈H₇Cl MW: 138.6 g/mol Boiling Pt: 189 °C Melting Pt: –63 °C Density: 1.091 Flash Pt: 65 °C (149 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000567 CAS Number: 2039-87-4 EINECS: 218-026-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...