Trifluralin
Supplier: LGC Standards PROMOCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2,6-Dinitro-N,N-dipropyl-4-trifluoromethylaniline
, α,α,α-Trifluoro-2,6-dinitro-N,N-dipropyl-p-toluidine
, N,N-Dipropyl-2,6-dinitro-4-trifluoromethylaniline
PROONMIAP1408EA
367
GBP
PROONMIAP1408
Trifluralin
Trifluralin
Formula:
C₁₃H₁₆F₃N₃O₄ MW: 335.28 g/mol Boiling Pt: 369 °C (1013 hPa) Melting Pt: 48 °C Density: 1.294 g/cm³ (20 °C) Flash Pt: 100 °C (closed cup) Storage Temperature: Ambient |
CAS Number:
1582-09-8 EINECS: 216-428-8 UN: 2811 ADR: 6.1,III Merck Index: 12,09815 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...