Congo red, Sigma-Aldrich®

Supplier: Merck Millipore Calbiochem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Sodium 3,3'-([1,1'-biphenyl]-4,4'-diylbis(diazene-2,1-diyl))bis(4-aminonaphthalene-1-sulphonate)Direct Red 28Disodium 3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalene-1-sulphonate)CI 22120

234610-500EA 100 GBP
234610-500
Congo red, Sigma-Aldrich®
Congo Red
Formula: C₃₂H₂₂N₆Na₂O₆S₂
MW: 696.67 g/mol
Melting Pt: > 360 °C
Storage Temperature: Ambient
MDL Number: MFCD00004028
CAS Number: 573-58-0
EINECS: 209-358-4
Merck Index: 12,02562

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR