Styrene-[D₈] (98% D) stabilised
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Styrene-[²H₈]
, Phenylethylene-D8
, Phenylethene-D8
, Vinylbenzene-D8
Stabilisation:
Stabilised with 4-tert-butylpyrocatechol (TBC)
ampouled under argon
43667.03EA
101
GBP
43667.03
43667.06
Styrene-[D₈] (98% D) stabilised
Styrene-[D8]
Formula:
C₆D₅CDCD₂ MW: 112.09 g/mol Boiling Pt: 50…51 °C (25 mmHg) Melting Pt: –31 °C Density: 0.979 Flash Pt: 31 °C (88 °F) |
MDL Number:
MFCD00044231 CAS Number: 19361-62-7 EINECS: 242-995-6 UN: 2055 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...