Styrene-[D₈]
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Styrene-[²H₈]
, Phenylethylene-D8
, Phenylethene-D8
, Vinylbenzene-D8
M-Clarity™ quality level = MQ200
8.15061.0010EA
1630
GBP
8.15061.0010
Styrene-[D₈]
Styrene-[D8]
Formula:
C₆D₅CDCD₂ MW: 112.09 g/mol Boiling Pt: 145 °C (1013 hPa) Melting Pt: –31 °C Density: 0.97 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00044231 CAS Number: 19361-62-7 EINECS: 242-995-6 UN: 2055 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...