Methyl bromoacetate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ200
SIAL157910-25GEA
38.8
GBP
SIAL157910-25G
SIAL157910-100G
Methyl bromoacetate, Sigma-Aldrich®
Methyl bromoacetate
Formula:
BrCH₂COOCH₃ MW: 152.98 g/mol Boiling Pt: 145 °C (1013 hPa) Melting Pt: –50 °C Density: 1.635 g/cm³ (20 °C) Flash Pt: 60 °C |
MDL Number:
MFCD00000189 CAS Number: 96-32-2 EINECS: 202-499-2 UN: 2643 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...