1-Chloroethyl chloroformate
Supplier: Molekula
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MOLEM83728178EA
0
GBP
MOLEM83728178
1-Chloroethyl chloroformate
1-Chloroethyl chloroformate
Formula:
C₃H₄Cl₂O₂ MW: 142.97 g/mol Boiling Pt: 118…119 °C (1013 hPa) Melting Pt: –65 °C Density: 1.31 g/cm³ (20 °C) Flash Pt: 40 °C |
MDL Number:
MFCD00000647 CAS Number: 50893-53-3 EINECS: 256-834-2 UN: 2742 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...