3-Nitrophthalic acid
Supplier: Molekula
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
MOLE64133332-100GEA
0
GBP
MOLE64133332-100G
3-Nitrophthalic acid
3-Nitrophthalic acid
Formula:
C₈H₅NO₆ MW: 211.13 g/mol Boiling Pt: 441 °C (1013 hPa) Melting Pt: 210…215 °C |
MDL Number:
MFCD00007138 CAS Number: 603-11-2 EINECS: 210-030-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...