Methyl bromoacetate, Sigma-Aldrich®
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
M-Clarity™ quality level = MQ200
8.20196.9999
Methyl bromoacetate, Sigma-Aldrich®
Methyl bromoacetate
Formula:
BrCH₂COOCH₃ MW: 152.98 g/mol Boiling Pt: 145 °C (1013 hPa) Melting Pt: –50 °C Density: 1.635 g/cm³ (20 °C) Flash Pt: 60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000189 CAS Number: 96-32-2 EINECS: 202-499-2 UN: 2643 ADR: 6.1,II |
Specification Test Results
Solubility | (20 °C) practically insoluble |
Molar Mass | 152.98 g/mol |
Boiling Point | 144 - 146 °C (1013 hPa) |
Vapor Pressure | 18 hPa (50 °C) |
Flash Point | 64 °C |
Refractive Index | 1.4586 (20 °C, 589 nm) |
Density | 1.65 g/cm3 (20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...