5-Nitrobenzimidazole, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ200
8.18257.0100EA
93.9
GBP
8.18257.0100
8.18257.9999
5-Nitrobenzimidazole, Sigma-Aldrich®
5-Nitrobenzimidazole
Formula:
C₇H₅N₃O₂ MW: 163.14 g/mol Melting Pt: 200…203 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005604 CAS Number: 94-52-0 EINECS: 202-341-2 UN: 2811 ADR: 6.1,II |
Specification Test Results
Solubility | 0.5 g/l (25 °C) |
Melting Point | 204 - 208 °C |
Molar Mass | 163.14 g/mol |
Bulk Density | 270 kg/m3 |
pH | 6 (10 g/l, H2O, 20 °C) (slurry) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...