Ethyl chloroacetate for synthesis, Sigma-Aldrich®
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
|
M-Clarity™ quality level = MQ200
MERC8.00413.9999
Ethyl chloroacetate for synthesis, Sigma-Aldrich®
Ethyl chloroacetate
Formula:
C₄H₇ClO₂ MW: 122.55 g/mol Boiling Pt: 144 °C (1013 hPa) Melting Pt: –26 °C Density: 1.15 g/cm³ (20 °C) Flash Pt: 56 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000932 CAS Number: 105-39-5 EINECS: 203-294-0 ADR: 6.1 (3),II Merck Index: 13,03818 |
Specification Test Results
Solubility | 12.3 g/l (20 °C) |
Melting Point | -26 °C |
Molar Mass | 122.55 g/mol |
Boiling Point | 144 °C (1013 hPa) |
Vapor Pressure | 4.8 hPa (20 °C) |
Flash Point | 54 °C |
Refractive Index | 1.4205 (20 °C, 589 nm) |
Explosion Limit | 2.6 %(V) |
Density | 1.15 g/cm3 (20 °C) |
Ignition Point | 452 °C |
Evaporation number | 0.23 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...