3-Hydroxy-5-methylisoxazole

Supplier: EHRENSTORFER GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Hymexazol

EHERC14270000EA 288 GBP
EHERC14270000
3-Hydroxy-5-methylisoxazole
3-Hydroxy-5-methylisoxazole
Formula: C₄H₅NO₂
MW: 99.09 g/mol
MDL Number: MFCD00144468
CAS Number: 10004-44-1
EINECS: 233-000-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR