5-Fluoro-1H-indazole ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
H26033.03EA
141
GBP
H26033.03
5-Fluoro-1H-indazole ≥98%
5-Fluoro-1H-indazole
Formula:
C₇H₅FN₂ MW: 136.13 g/mol Melting Pt: 122…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD04972877 CAS Number: 348-26-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...