Isovaleryl chloride ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
B24002.36EA
122
GBP
B24002.36
B24002.14
B24002.22
Isovaleryl chloride ≥98%
Isovaleryl chloride
Formula:
C₅H₉ClO MW: 120.58 g/mol Boiling Pt: 115…117 °C Density: 0.989 Flash Pt: 27 °C (80 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000738 CAS Number: 108-12-3 EINECS: 203-552-0 UN: 2920 ADR: 8,II Merck Index: 13,05253 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...