cis-4-Decen-1-ol ≥97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Cis-4-decen-1-ol
, cis-Dec-4-en-1-ol
A18311.14EA
0
GBP
A18311.14
cis-4-Decen-1-ol ≥97%
cis-4-Decen-1-ol
Formula:
C₁₀H₂₀O MW: 156.27 g/mol Boiling Pt: 231…232 °C Flash Pt: 105 °C (221 °F) |
MDL Number:
MFCD00015568 CAS Number: 57074-37-0 EINECS: 260-548-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...