United Kingdom
Order Entry
Recombinant Human IL10 (from Sf9 cells), His-tag
  ABNOP8316.10UG
 :  Abnova
 :  
Recombinant Human IL10 (from Sf9 cells), His-tag
  ABNOP8316.10UG
 :  Abnova
 :  
 
£261.00
   
  1 * 10 µG

 

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Insect cells (Sf9)
  • Species:
    Human
  • Environmentally Preferable:
  • Tag sequence:
    His-tag
  • Gene ID:
    P22301
  • Protein/peptide name:
    IL10
  • Sequence:
    19a.a.-178a.a.