- Antibody type:Primary
- Antigen name:C3orf39
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:221-320/580
- Form:Liquid
- Gene ID:84892
- Antigen synonyms:AGO61|Uncharacterized glycosyltransferase AGO61 precursor|FLJ14566|GTDC2_HUMAN|Chromosome 3 open reading frame 39
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human C3orf39
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
C3orf39, also known as AGO61, is a 580 amino acid secreted protein belonging to the glycosyltransferase 61 family. C3orf39 is encoded by a gene that maps to human chromosome 3p25.1. Chromosome 3 is made up of approximately 214 million bases encoding over 1100 genes. Notably, there is a chemokine receptor gene cluster and a variety of human cancer related loci on chromosome 3. Particular regions of the chromosome 3 short arm are deleted in many types of cancer cells. Key tumour suppressing genes on chromosome 3 encode apoptosis mediator RASSF1, cell migration regulator HYAL1 and angiogenesis suppressor SEMA3B. Marfan Syndrome, porphyria, von Hippel-Lindau syndrome, osteogenesis imperfecta and Charcot-Marie-Tooth disease are a few of the numerous genetic diseases associated with chromosome 3.
Type: Primary
Antigen: C3orf39
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 221-320/580
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat