Anti-SCOT2 Rabbit Polyclonal Antibody (FITC)
BOSSBS-15589R-FITC
: Bioss
- Antibody type:Primary
- Antigen name:SCOT2
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:101-200/517
- Form:Liquid
- Antigen synonyms:3-oxoacid CoA-transferase 2A|Testis-specic succinyl-CoA:3-oxoacid CoA-transferase|SCOT2_HUMAN|SCOT-t|Succinyl-CoA:3-ketoacid coenzyme A transferase 2, mitochondrial
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:52 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human SCOT2
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
OXCT2 is a testis-specific succinyl-CoA:3-oxoacid CoA transferase (EC 2,8,3,5), which catalyses the reversible transfer of CoA from succinyl-CoA to acetoacetate in the first step of ketone body utilisation. See also OXCT1 (MIM 601424).
Type: Primary
Antigen: SCOT2
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 101-200/517
Host: Rabbit
Isotype: IgG
Reactivity: Human