Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

You Searched For:

Microcarriers

8  results were found
https://uk.vwr.com/stibo/smallweb/std.lang.all/95/30/18989530.jpg

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"8"
  List View Easy View
Sort by:
 
 
 
 

Supplier:  GE Healthcare - Life Sciences
Description:   Cytodex 3, formed by chemically coupling a thin layer of denatured collagen to the cross-linked dextran matrix, is the microcarrier of choice for cell...
Catalog Number: (17-1271-01)

Supplier:  GE Healthcare - Life Sciences
Description:   Cytopore 2 macroporous microcarriers are principally designed for use in suspension culture systems for growth of adherent diploid cells requiering a ...
UOM:  1 * 20 g
Catalog Number: (17-0911-01)

Supplier:  GE Healthcare - Life Sciences
Description:   Cytopore1 macroporous microcarriers are principally designed for use in suspension culture systems for growth of adherent recombinant Chinese Hamster ...
UOM:  1 * 20 g
Supplier:  GE Healthcare - Life Sciences
Description:   Cytodex 1 is particularly suitable for most established cell lines and for production of viruses or cell products from cultures of primary cells and n...
Supplier:  GE Healthcare - Life Sciences
Description:   The microcarrier of choice for cells that are difficult to culture in vitro - particularly for cells with an epithelial-like morphology.
Supplier:  GE Healthcare - Life Sciences
Description:   Sterilized Cytodex 1 Gamma is a general-purpose microcarrier particularly suitable for most established cell lines and for production of viruses or ce...
Supplier:  Corning Life Science
Description:   The new Corning Dissolvable Microcarriers are designed to simplify the cell harvest, separation, and concentration processes. All Microcarriers arrive...
Supplier:  Corning Life Science
Description:   Corning® Microcarriers are designed to simplify bioprocessing scale up. The Microcarriers are sterile, ready to use, and available in closed systems p...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on 0800 22 33 44.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on 0800 22 33 44
Additional Documentation may be needed to purchase this item. A VWR representative will contact you if needed.
Additional Documentation may be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service on 0800 22 33 44.
1 - 8  of 8