4-Bromo-2-(piperidin-1-ylsulphonyl)anisole 97%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-((5-Bromo-2-methoxyphenyl)sulphonyl)piperidine
APOSOR9412-1GEA
107
GBP
APOSOR9412-1G
4-Bromo-2-(piperidin-1-ylsulphonyl)anisole 97%
4-Bromo-2-(piperidin-1-ylsulphonyl)anisole
Formula:
C₁₂H₁₆BrNO₃S MW: 334.23 g/mol Melting Pt: 95…99 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02043682 CAS Number: 295360-83-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...