Propargyl ether
APOSOR14509-10G
:
- Pack type:Bottle
- Pk:10 g
- Seq:3236
- Environmentally Preferable:
: Dipropargylether
:
:94.11 g/mol
:55…57 °C/65mmHg
:0.914 g/cm³ (25 °C)
:24 °C
:3271
:3,III
:GHS02
:GHS07
:Refrigerator
:6921-27-3
:C₆H₆O
:MFCD00048108