6-Bromo-8-nitro[1,2,4]triazolo[4,3-a]pyridine
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
APOSOR11591-1GEA
0
GBP
APOSOR11591-1G
6-Bromo-8-nitro[1,2,4]triazolo[4,3-a]pyridine
6-Bromo-8-nitro-[1,2,4]triazolo[4,3-a]pyridine
Formula:
C₆H₃BrN₄O₂ Storage Temperature: Ambient |
MDL Number:
MFCD09801055 CAS Number: 951884-20-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...