5-Bromo-4-o-tolylpyrimidine 98%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
5-Bromo-4-(2-methylphenyl)pyrimidine
APOSOR11554-1GEA
0
GBP
APOSOR11554-1G
APOSOR11554-5G
5-Bromo-4-o-tolylpyrimidine 98%
5-Bromo-4-o-tolylpyrimidine
Formula:
C₁₂H₁₁BrN₂ Storage Temperature: Ambient |
MDL Number:
MFCD09475895 CAS Number: 941294-34-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...