5-Carboxyfluorescein, Sigma-Aldrich®
Supplier: SIGMA-ALDRICH MICROSCOPY
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Spiro(isobenzofuran-1(3H),9'-(9H)xanthene)-5-carboxylic acid, 3',6'-dihydroxy-3-oxo-
, 5-Carboxyfluorescein hydrate
, 3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-5-carboxylic acid
, 5-FAM
C0537-50MGEA
123
GBP
C0537-50MG
5-Carboxyfluorescein, Sigma-Aldrich®
5-Carboxyfluorescein
5-Carboxyfluorescein has been used as an internal marker in capillary isotachophoresis to quantify the charge-based low density lipoprotein subfractions.
Formula:
C₂₁H₁₂O₇.xH₂O MW: 376.32 g/mol (anhy) Storage Temperature: Refrigerator |
MDL Number:
MFCD00036874 CAS Number: 76823-03-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...