IDT-2BR, Organic Photovoltaics (OPV) reagent

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

H66830.MCEA 442 GBP
H66830.MC H66830.ME H66830.MD
IDT-2BR, Organic Photovoltaics (OPV) reagent
IDT-2BR
Formula: C₈₈H₈₈N₆O₂S₈
MW: 1518.20 g/mol
CAS Number: 2042521-91-3

Order Now

Specification Test Results

Appearance (Color) Black
Form Flake or threadlike
Bandgap energy 1,69 ± 0,02 eV

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR