BTP-4Cl, Organic Photovoltaics (OPV) reagent

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: TTPTTI-4C

H66315.MCEA 485.68 GBP
H66315.MC H66315.ME H66315.MD
BTP-4Cl, Organic Photovoltaics (OPV) reagent
BTP-4Cl
Formula: C₈₂H₈₆Cl₄N₈O₂S₅
MW: 1517.75 g/mol
CAS Number: 2414918-25-3

Order Now

Specification Test Results

Appearance (Color) Black
Form Flake or threadlike
Bandgap energy 1,36 ± 0,02 eV

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR