PffBT4T-2OD (PCE11), Organic Photovoltaics (OPV) reagent
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H66126.MCEA
512
GBP
H66126.MC
H66126.MD
H66126.ME
PffBT4T-2OD (PCE11), Organic Photovoltaics (OPV) reagent
PffBT4T-2OD (PCE11)
Formula:
(C₆₂H₈₈N₂S₅F₂)n |
CAS Number:
1644164-62-4 |
Specification Test Results
Appearance (Color) | Black |
Form | Flake or threadlike |
Bandgap energy | 1,63 ± 0,02 eV |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...