Aqueous mounting medium, Fluoroshield™ with PI, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Fluoroshield™
F5932-20MLEA 92.8 GBP
F5932-20ML
Aqueous mounting medium, Fluoroshield™ with PI, Sigma-Aldrich®
Stains Microscopy and staining reagents
Fluoroshield™ with PI is an aqueous mounting medium for preserving fluorescence of tissue and cell smears.

  • Storage temperature: 2 - 8 °C

This mounting medium is fortified with propidium iodide (PI) which is a counterstain for DNA. For use in in situ hybridization techniques or other methods where fluorescence of DNA staining is required. PI excites at 535 nm and emits at 615 nm, producing a red fluorescence. RNA is also stained with PI.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR