COV-Hygien Xpress on-site detection kit, BioMire

Supplier: CHROMagar
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BioMire
SARSSTP05EA 0 GBP
SARSSTP05 SARSSBK25
COV-Hygien Xpress on-site detection kit, BioMire
Clinical Diagnostic Systems Clinical Reagents and Kits
The COV-Hygien Xpress detection kit provides rapid, ready to use technology to detect SARS CoV-2 on surfaces and liquids.

  • Fast results: Detection results in under 15 minutes
  • Everything you need: Do it yourself without the need to transport the sample to a lab
  • Easy to use: No technical training nor instrument or equipment required; easy as water quality test strips
  • Specific: Detects SARS CoV-2 proteins: Minimises risk of false positives
  • Economical: Inexpensive compared to PCR techniques
  • A single test that can be used on surfaces or liquids

Benefits:
- Reassures the public that surfaces have been tested and sanitized for live viruses
- Improves sanitation procedures by routine testing for potential virus deposits
- Checks liquids for the presence of SARS CoV-2
- Immediately sanitises after on-site positive test and prevents further exposure to virus
- Increases test and monitoring data to support your procedures

Caution: NOT for use with samples from human origin. For environmental testing only.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR