Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Size exclusion chromatography columns, ProSEC™ 300S

Supplier: Agilent Technologies
ProSec™ 300 S
ProSEC™ 300S is a silica-based material specifically designed for the analysis of proteins by aqueous size exclusion chromatography. With a proprietary surface modification to remove acidic silanols and reduce interaction effects, the ProSEC packing material shows minimal non-size exclusion effects for a wide range of protein types. The packing material has a nominal 300 Å pore size suitable for a wide range of globular proteins and a 5 µm particle size that gives a high efficiency for improved resolution, making the new ProSEC™ 300S an ideal choice for all protein analyses.

  • The high surface stability ensures that the column is suitable for use in buffers with a pH range between 2,0 and 7,5
  • A 300 Å pore size suited to the analysis of a wide range of therapeutic and biologically active globular proteins
  • High mechanical stability makes for excellent column lifetimes and gives high sample throughput




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
I-Ø×L Pk VWR Catalog Number Quantity
4,6×250 1 554-1692
7,5×300 1 554-1689
7,5×600 1 554-1690

Guard Columns
I-Ø×L Pk VWR Catalog Number Quantity
4,6×50 1 554-1691
7,5×50 1 554-1688
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty