n-Heptane
Supplier: LGC Standards PROMOCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
PROOIRMM-441EA
966
GBP
PROOIRMM-441
XXXXIRMM-441
n-Heptane
n-Heptane
Formula:
H₃C(CH₂)₅CH₃ MW: 100.2 g/mol Boiling Pt: 98.4 °C (1013 hPa) Melting Pt: –90.6 °C Density: 0.6838 g/cm³ (20 °C) Flash Pt: –4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009544 CAS Number: 142-82-5 EINECS: 205-563-8 UN: 1206 ADR: 3,II Merck Index: 13,04679 |
![]() |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...