Formalin neutral sodium salt, Gurr™ histological fixative, buffered pH 7.0 (25 °C)
Supplier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
351638KEA
265
GBP
351638K
Formalin neutral sodium salt, Gurr™ histological fixative, buffered pH 7.0 (25 °C)
Formalin neutral sodium salt
Formula:
CH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00003274 CAS Number: 50-00-0 EINECS: 200-001-8 UN: 3334 ADR: 9,III |
Specification Test Results
pH (25°C) | 6.9 - 7.1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...