Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Viscometer with falling ball

Supplier: Fungilab
The VISCO BALL measures the viscosity of transparent Newtonian liquids and gases and is particularly suited to low viscosity materials. It has applications in research and quality control for a wide range of industries including food, cosmetics, petroleum detergents and paints. According to the Hoppler principle, the time is measured for a ball to travel a reference distance through the sample liquid in a cylindrical tube when inclined at 80°, and this measurement correlates to a dynamic viscosity. When the temperature is controlled precisely the viscosity result from this type of viscometer meets DIN 53015 and ISO 12058 standards and has very good accuracy. The VISCO BALL can be also be inclined and secured at angles other than 80° to meet different individual user requirements.

  • High accuracy with reproducibility better than 0,5 % and comparability better than 1,0 %
  • Extended re-calibration periods
  • A ball is available for gases (option)

Suitable for applications in research and quality control in food, cosmetics, petroleum detergents and paintings.

According to the Hoppler principle, the time is measured for a ball to travel a reference distance through the sample liquid in a cylindrical tube when inclined at 80° and this measurement correlates to a dynamic viscosity.

The tube can be turned up and the measurement repeated.

Certifications: DIN 53015 - ISO 12058



 

ORDER

Expand All / Collapse All
Description Range Pk VWR Catalog Number Quantity
Viscometer with falling ball 0,6 − 105 cP 1 645-0005
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Pk VWR Catalog Number Quantity
Glass thermometer (24…50 °C ; 0,1 °C) 1 FUNGV91002
Glass thermometer (49…76 °C ; 0,1 °C) 1 FUNGV91003
Glass thermometer (74…101 °C ; 0,1 °C) 1 FUNGV91004
Glass thermometer (99…126 °C ; 0,1 °C) 1 FUNGV91005
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty