Sodium hydroxide solution 32% (max. 0.002% K), EMSURE® for analysis, Supelco®
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Caustic soda
M-Clarity™ quality level = MQ300
1.05500.2500EA
285
GBP
1.05500.2500
Sodium hydroxide solution 32% (max. 0.002% K), EMSURE® for analysis, Supelco®
Sodium hydroxide
Formula:
NaOH MW: 40 g/mol Melting Pt: 9 °C Density: 1.35 g/cm3 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 UN: 1824 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...