2-Butyne-1,4-diol unstabilised, GPR RECTAPUR®

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

85747.460EA 0 GBP
85747.460 85747.260
2-Butyne-1,4-diol unstabilised, GPR RECTAPUR®
2-Butyne-1,4-diol
Formula: C₄H₆O₂
MW: 86.09 g/mol
Boiling Pt: 238 °C (1013 hPa)
Melting Pt: 55…58 °C
Density: 1.1001 g/cm³ (20 °C)
Flash Pt: 142 °C
Storage Temperature: Ambient
MDL Number: MFCD00002915
CAS Number: 110-65-6
EINECS: 203-788-6
UN: 2716
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow to beige crystals or flakes
Assay (GC) ≥99.0 %
Melting range (lower value) ≥54 °C
Melting range (upper value) ≤57 °C
Identity (IR) passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR